3,6-Dichloro-4-methoxypyridazine 97%

BD164349-5G BD164349-1G
BLDPBD164349-5GEA 44.1 EUR
BLDPBD164349-5G BLDPBD164349-1G
3,6-Dichloro-4-methoxypyridazine 97%
3,6-Dichloro-4-methoxypyridazine

Storage condition: Inert atmosphere, 2-8°C

Formula: C₅H₄Cl₂N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD00477252
CAS Number: 70952-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR