3-Bromo-2-methylbenzotrifluoride 98%
|
|
BD164369-10G
BD164369-100G
BLDPBD164369-10GEA
52.5
EUR
BLDPBD164369-10G
BLDPBD164369-100G
3-Bromo-2-methylbenzotrifluoride 98%
3-Bromo-2-methylbenzotrifluoride
Storage condition: Sealed in dry, Room Temperature
Formula:
C₈H₆BrF₃ Storage Temperature: Ambient |
MDL Number:
MFCD04038231 CAS Number: 69902-83-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...