Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate 97%

BD164663-1G BD164663-10G
BLDPBD164663-1GEA 23.1 EUR
BLDPBD164663-1G BLDPBD164663-10G
Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate 97%
Methyl 1,4-Dioxaspiro[4.5]decane-8-carboxylate

Storage condition: Inert atmosphere, Room Temperature

Formula: C₁₀H₁₆O₄
MW: 200.23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13195278
CAS Number: 26845-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR