5-Bromo-2-cyclopropylpyridine 98%

BD164736-250MG BD164736-1G
BLDPBD164736-250MGEA 92.4 EUR
BLDPBD164736-250MG BLDPBD164736-1G
5-Bromo-2-cyclopropylpyridine 98%
5-Bromo-2-cyclopropylpyridine

Storage condition: Sealed in dry, Room Temperature

Formula: C₈H₈BrN
Storage Temperature: Ambient
MDL Number: MFCD12033369
CAS Number: 579475-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR