4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-isoindol-1-one 98%
|
|
BD164779-100MG
BD164779-250MG
BLDPBD164779-100MGEA
41
EUR
BLDPBD164779-100MG
BLDPBD164779-250MG
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-isoindol-1-one 98%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1H-isoindol-1-one
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₄H₁₈BNO₃ Storage Temperature: Refrigerator |
MDL Number:
MFCD11617913 CAS Number: 765948-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...