4-Amino-2-bromo-6-methylpyridine 97%
|
|
BD164783-1G
BD164783-250MG
BLDPBD164783-1GEA
81.9
EUR
BLDPBD164783-1G
BLDPBD164783-250MG
4-Amino-2-bromo-6-methylpyridine 97%
4-Amino-2-bromo-6-methylpyridine
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₆H₇BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD12400760 CAS Number: 79055-59-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...