Boc-(R)-3-amino-4-(3-methylphenyl)butyric acid 98%

BD16481-100MG BD16481-250MG
BLDPBD16481-100MGEA 88.2 EUR
BLDPBD16481-100MG BLDPBD16481-250MG
Boc-(R)-3-amino-4-(3-methylphenyl)butyric acid 98%
Boc-(R)-3-amino-4-(3-methylphenyl)butyric acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₆H₂₃NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD01860925
CAS Number: 269398-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR