2-(4-Morpholino)-5-pyrimidineboronic acid pinacol ester 97%

BD165105-250MG BD165105-25G
BLDPBD165105-250MGEA 45.2 EUR
BLDPBD165105-250MG BLDPBD165105-25G
2-(4-Morpholino)-5-pyrimidineboronic acid pinacol ester 97%
2-(4-Morpholino)-5-pyrimidineboronic acid pinacol ester

Storage condition: Inert atmosphere, 2-8°C

Formula: C₁₄H₂₂BN₃O₃
MW: 291.1538 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368246
CAS Number: 957198-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR