4-Bromo-6-methylpyrimidine 95%
|
|
BD165473-100MG
BD165473-5G
BLDPBD165473-100MGEA
33.6
EUR
BLDPBD165473-100MG
BLDPBD165473-5G
4-Bromo-6-methylpyrimidine 95%
4-Bromo-6-methylpyrimidine
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₅H₅BrN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00234065 CAS Number: 69543-98-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...