1-Benzhydryl-3-azetidinyl methanesulphonate 97%

BD16711-1G BD16711-100G
BLDPBD16711-1GEA 7.4 EUR
BLDPBD16711-1G BLDPBD16711-100G
1-Benzhydryl-3-azetidinyl methanesulphonate 97%
1-Benzhydryl-3-azetidinyl methanesulphonate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₇H₁₉NO₃S
MW: 317.40 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00159216
CAS Number: 33301-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR