2-Iodoresorcinol 98%

BD167228-5G BD167228-100MG
BLDPBD167228-5GEA 374 EUR
BLDPBD167228-5G BLDPBD167228-100MG
2-Iodoresorcinol 98%
2-Iodoresorcinol

Storage condition: Keep in dark place, Inert atmosphere, 2-8°C

Formula: C₆H₅IO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09701440
CAS Number: 41046-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR