2-Amino-5-bromo-3-(trifluoromethyl)pyridine 98%

BD167598-1G BD167598-100G
BLDPBD167598-1GEA 24.2 EUR
BLDPBD167598-1G BLDPBD167598-100G
2-Amino-5-bromo-3-(trifluoromethyl)pyridine 98%
2-Amino-5-bromo-3-(trifluoromethyl)pyridine

Storage condition: Keep in dark place, Inert atmosphere, Room temperature

Formula: C₆H₄BrF₃N₂
MW: 241.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10758088
CAS Number: 79456-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR