5-Bromo-3-chloropyridine-2-carboxylic acid 98%
|
|
BD167806-25G
BD167806-10G
BLDPBD167806-25GEA
212
EUR
BLDPBD167806-25G
BLDPBD167806-10G
5-Bromo-3-chloropyridine-2-carboxylic acid 98%
5-Bromo-3-chloropyridine-2-carboxylic acid
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₆H₃BrClNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD12827552 CAS Number: 1189513-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...