4-Fluoro-1H-indole-7-carbonitrile 97%

BD167931-250MG BD167931-100MG
BLDPBD167931-250MGEA 105 EUR
BLDPBD167931-250MG BLDPBD167931-100MG
4-Fluoro-1H-indole-7-carbonitrile 97%
4-Fluoro-1H-indole-7-carbonitrile

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₅FN₂
Storage Temperature: Ambient
MDL Number: MFCD11845597
CAS Number: 313337-33-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR