5-Amino-2-methylthiazole-4-carboxylic acid ethyl ester 98%

BD167933-100MG BD167933-5G
BLDPBD167933-100MGEA 23.1 EUR
BLDPBD167933-100MG BLDPBD167933-5G
5-Amino-2-methylthiazole-4-carboxylic acid ethyl ester 98%
5-Amino-2-methylthiazole-4-carboxylic acid ethyl ester

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₇H₁₀N₂O₂S
MW: 186.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07357418
CAS Number: 31785-05-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR