Tris(4-bromophenyl)amine 98%
|
|
BD16796-25G
BD16796-10G
BLDPBD16796-25GEA
26.3
EUR
BLDPBD16796-25G
BLDPBD16796-10G
Tris(4-bromophenyl)amine 98%
Tris(4-bromophenyl)amine
Storage condition: Sealed in dry, Room Temperature
Formula:
C₁₈H₁₂Br₃N MW: 482.01 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00009665 CAS Number: 4316-58-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...