2-Bromo-5-chloro-3-fluoropyridine 98%

BD167975-5G BD167975-1G BD167975-100G
BLDPBD167975-5GEA 23.1 EUR
BLDPBD167975-5G BLDPBD167975-1G BLDPBD167975-100G
2-Bromo-5-chloro-3-fluoropyridine 98%
2-Bromo-5-chloro-3-fluoropyridine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₅H₂BrClFN
Storage Temperature: Ambient
MDL Number: MFCD11044268
CAS Number: 514797-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR