2,5-Diphenyl-p-benzoquinone 99%

BD16800-25G BD16800-5G
BLDPBD16800-25GEA 153 EUR
BLDPBD16800-25G BLDPBD16800-5G
2,5-Diphenyl-p-benzoquinone 99%
2,5-Diphenyl-p-benzoquinone

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₈H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00001600
CAS Number: 844-51-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR