2-Chloro-5-nitroisonicotinic acid 98%
|
|
BD168527-1G
BD168527-25G
BLDPBD168527-1GEA
13.7
EUR
BLDPBD168527-1G
BLDPBD168527-25G
2-Chloro-5-nitroisonicotinic acid 98%
2-Chloro-5-nitroisonicotinic acid
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₆H₃ClN₂O₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD14584664 CAS Number: 907545-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...