5-Bromo-4,6-dimethylpyrimidine 97%

BD168591-5G BD168591-25G
BLDPBD168591-5GEA 236 EUR
BLDPBD168591-5G BLDPBD168591-25G
5-Bromo-4,6-dimethylpyrimidine 97%
5-Bromo-4,6-dimethylpyrimidine

Storage condition: Sealed in dry, 2-8°C

Formula: C₆H₇BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD08059561
CAS Number: 157335-97-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR