1,1'-Diheptyl-4,4'-bipyridinium dibromide 95%
|
|
BD1689-5G
BD1689-25G
BLDPBD1689-5GEA
92.4
EUR
BLDPBD1689-5G
BLDPBD1689-25G
1,1'-Diheptyl-4,4'-bipyridinium dibromide 95%
1,1'-Diheptyl-4,4'-bipyridinium dibromide
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₂₄H₃₈Br₂N₂ MW: 514.39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011997 CAS Number: 6159-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...