BOC-L-phenylalanine-N-hydroxysuccinimide ester 97%
|
|
BD16935-5G
BD16935-25G
BLDPBD16935-5GEA
16.8
EUR
BLDPBD16935-5G
BLDPBD16935-25G
BOC-L-phenylalanine-N-hydroxysuccinimide ester 97%
BOC-L-phenylalanine-N-hydroxysuccinimide ester
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Formula:
C₁₈H₂₂N₂O₆ Storage Temperature: Freezer |
MDL Number:
MFCD00037911 CAS Number: 3674-06-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...