N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic acid 97%

BD17017-1G BD17017-10G
BLDPBD17017-1GEA 10.5 EUR
BLDPBD17017-1G BLDPBD17017-10G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic acid 97%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₁₉H₁₇NO₆
Storage Temperature: Refrigerator
MDL Number: MFCD01318740
CAS Number: 136083-57-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR