2,6-Bis[(2-hydroxyethyl)amino]toluene 95%
|
|
BD17052-10G
BD17052-25G
BLDPBD17052-10GEA
24.2
EUR
BLDPBD17052-10G
BLDPBD17052-25G
2,6-Bis[(2-hydroxyethyl)amino]toluene 95%
2,6-Bis[(2-hydroxyethyl)amino]toluene
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₁₁H₁₈N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD08703311 CAS Number: 149330-25-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...