3-Aminopiperidine-2,6-dione hydrochloride 97%

BD170886-5G BD170886-100G
BLDPBD170886-5GEA 9.5 EUR
BLDPBD170886-5G BLDPBD170886-100G
3-Aminopiperidine-2,6-dione hydrochloride 97%
3-Aminopiperidine-2,6-dione hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C5H9ClN2O2
Storage Temperature: Ambient
MDL Number: MFCD11042437
CAS Number: 24666-56-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR