(1-Aminocyclopropyl)methanol hydrochloride 97%

BD170922-100MG BD170922-10G
BLDPBD170922-100MGEA 9.5 EUR
BLDPBD170922-100MG BLDPBD170922-10G
(1-Aminocyclopropyl)methanol hydrochloride 97%
(1-Aminocyclopropyl)methanol hydrochloride

Storage condition: Inert atmosphere, Room Temperature

Formula: C₄H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD09834330
CAS Number: 115652-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR