5-Bromo-2-methoxybenzonitrile 97%

BD171309-1G BD171309-250MG
BLDPBD171309-1GEA 13.7 EUR
BLDPBD171309-1G BLDPBD171309-250MG
5-Bromo-2-methoxybenzonitrile 97%
5-Bromo-2-methoxybenzonitrile

Storage condition: Inert atmosphere, 2-8°C

Formula: C₈H₆BrNO
MW: 212.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00143429
CAS Number: 144649-99-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR