4,4'-Bipiperidine 98%

BD171337-100MG BD171337-5G
BLDPBD171337-100MGEA 48.3 EUR
BLDPBD171337-100MG BLDPBD171337-5G
4,4'-Bipiperidine 98%
4,4'-Bipiperidine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₀H₂₀N₂
Storage Temperature: Ambient
MDL Number: MFCD01104315
CAS Number: 15336-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR