4-Cyanophenyl-trans-4-(4-n-pentylcyclohexyl)benzoate 97%

BD17150-25G BD17150-1G
BLDPBD17150-25GEA 53.6 EUR
BLDPBD17150-25G BLDPBD17150-1G
4-Cyanophenyl-trans-4-(4-n-pentylcyclohexyl)benzoate 97%
4-Cyanophenyl-trans-4-(4-n-pentylcyclohexyl)benzoate

Storage condition: Sealed in dry, Room Temperature

Formula: C₂₅H₂₉NO₂
Storage Temperature: Ambient
MDL Number: MFCD09750946
CAS Number: 72928-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR