2-Amino-6-chlorophenol 98%
|
|
BD172038-5G
BD172038-1G
BLDPBD172038-5GEA
109
EUR
BLDPBD172038-5G
BLDPBD172038-1G
2-Amino-6-chlorophenol 98%
2-Amino-6-chlorophenol
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₆H₆ClNO Storage Temperature: Ambient |
MDL Number:
MFCD07801210 CAS Number: 38191-33-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...