2,4,6-Collidine 98%

BD17226-10G BD17226-25G
BLDPBD17226-10GEA 6.3 EUR
BLDPBD17226-10G BLDPBD17226-25G
2,4,6-Collidine 98%
2,4,6-Collidine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₈H₁₁N
Storage Temperature: Ambient
MDL Number: MFCD00006338
CAS Number: 108-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR