[(4-Fluorophenyl)amino](oxo)acetic acid 98%

BD172835-1G BD172835-25G
BLDPBD172835-1GEA 32.6 EUR
BLDPBD172835-1G BLDPBD172835-25G
[(4-Fluorophenyl)amino](oxo)acetic acid 98%
[(4-Fluorophenyl)amino](oxo)acetic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₈H₆FNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD09864524
CAS Number: 69066-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR