Fmoc-N-methyl-L-leucine 98%

BD17351-10G BD17351-5G
BLDPBD17351-10GEA 55.7 EUR
BLDPBD17351-10G BLDPBD17351-5G
Fmoc-N-methyl-L-leucine 98%
Fmoc-N-methyl-L-leucine

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₂₂H₂₅NO₄
MW: 367.445 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151933
CAS Number: 103478-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR