(S)-3-Methyl-1-(2-(piperidin-1-yl)phenyl)butan-1-amine(S)-2-acetamidopentanedioate 95%

BD173570-25G BD173570-5G
BLDPBD173570-25GEA 81.9 EUR
BLDPBD173570-25G BLDPBD173570-5G
(S)-3-Methyl-1-(2-(piperidin-1-yl)phenyl)butan-1-amine(S)-2-acetamidopentanedioate 95%
(S)-3-Methyl-1-(2-(piperidin-1-yl)phenyl)butan-1-amine(S)-2-acetamidopentanedioate

Storage condition: Inert atmosphere, 2-8°C

Formula: C₂₃H₃₇N₃O₅
Storage Temperature: Refrigerator
MDL Number: MFCD09840998
CAS Number: 219921-94-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR