4-Chloro-2-(methylsulphanyl)pyrimidine-5-carbonitrile 98%
|
|
BD174164-1G
BD174164-10G
BLDPBD174164-1GEA
52.5
EUR
BLDPBD174164-1G
BLDPBD174164-10G
4-Chloro-2-(methylsulphanyl)pyrimidine-5-carbonitrile 98%
4-Chloro-2-(methylsulphanyl)pyrimidine-5-carbonitrile
Storage condition: Inert atmosphere, Store in freezer, under -20°C
Formula:
C₆H₄ClN₃S Storage Temperature: Freezer |
MDL Number:
MFCD00507261 CAS Number: 33089-15-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...