4-Bromo-6-fluoro-1H-indazole 95%

BD174321-100MG BD174321-1G
BLDPBD174321-100MGEA 11.6 EUR
BLDPBD174321-100MG BLDPBD174321-1G
4-Bromo-6-fluoro-1H-indazole 95%
4-Bromo-6-fluoro-1H-indazole

Storage condition: Sealed in dry, Room Temperature

Formula: C₇H₄BrFN₂
Storage Temperature: Ambient
MDL Number: MFCD07781448
CAS Number: 885520-35-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR