(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(6-chloro-1H-indol-3-yl)propanoic acid 97%

BD174338-5G BD174338-100MG
BLDPBD174338-5GEA 795 EUR
BLDPBD174338-5G BLDPBD174338-100MG
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(6-chloro-1H-indol-3-yl)propanoic acid 97%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(6-chloro-1H-indol-3-yl)propanoic acid

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₆H₂₁ClN₂O₄
Storage Temperature: Refrigerator
MDL Number: MFCD16883068
CAS Number: 908847-42-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR