2,5-Dimethyl-2,5-hexanediol 98%
|
|
BD17502-25G
BD17502-100G
BLDPBD17502-25GEA
7.4
EUR
BLDPBD17502-25G
BLDPBD17502-100G
2,5-Dimethyl-2,5-hexanediol 98%
2,5-Dimethyl-2,5-hexanediol
Storage condition: Keep in dark place, Sealed in dry, Room Temperature
Formula:
C₈H₁₈O₂ Storage Temperature: Ambient |
MDL Number:
MFCD00004473 CAS Number: 110-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...