(4-Methylphenoxy)acetyl chloride 97%

BD175379-250MG BD175379-5G
BLDPBD175379-250MGEA 27.3 EUR
BLDPBD175379-250MG BLDPBD175379-5G
(4-Methylphenoxy)acetyl chloride 97%
(4-Methylphenoxy)acetyl chloride

Storage condition: Inert atmosphere, 2-8°C

Formula: C₉H₉ClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD00235216
CAS Number: 15516-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR