Fmoc-D-glutamine 98%

BD17590-25G BD17590-100G
BLDPBD17590-25GEA 139 EUR
BLDPBD17590-25G BLDPBD17590-100G
Fmoc-D-glutamine 98%
Fmoc-D-glutamine

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₀H₂₀N₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00062959
CAS Number: 112898-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR