4-(Dimethylamino)butan-2-one 95%

BD179060-1G BD179060-25G
BLDPBD179060-1GEA 50.4 EUR
BLDPBD179060-1G BLDPBD179060-25G
4-(Dimethylamino)butan-2-one 95%
4-(Dimethylamino)butan-2-one

Storage condition: Inert atmosphere, Room Temperature

Formula: C₆H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD00457365
CAS Number: 2543-57-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR