Diethyl 1H-indole-2,5-dicarboxylate 95%

BD17909-100MG BD17909-1G
BLDPBD17909-100MGEA 36.8 EUR
BLDPBD17909-100MG BLDPBD17909-1G
Diethyl 1H-indole-2,5-dicarboxylate 95%
Diethyl 1H-indole-2,5-dicarboxylate

Storage condition: Sealed in dry, Room Temperature

Formula: C₁₄H₁₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD03094964
CAS Number: 127221-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR