Fmoc-Glu(OAll)-OH 98%

BD18076-100G BD18076-25G
BLDPBD18076-100GEA 574 EUR
BLDPBD18076-100G BLDPBD18076-25G
Fmoc-Glu(OAll)-OH 98%
Fmoc-Glu(OAll)-OH

Storage condition: Sealed in dry, 2-8°C

Formula: C₂₃H₂₃NO₆
MW: 409.438 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00190879
CAS Number: 133464-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR