L-Valine-tert-butyl ester hydrochloride 95%
|
|
BD18130-25G
BD18130-1G
BD18130-5G
BLDPBD18130-25GEA
31.5
EUR
BLDPBD18130-25G
BLDPBD18130-1G
BLDPBD18130-5G
L-Valine-tert-butyl ester hydrochloride 95%
L-Valine-tert-butyl ester hydrochloride
Storage condition: Inert atmosphere, Room Temperature
Formula:
C₉H₁₉NO₂·HCl MW: 209.7 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00034863 CAS Number: 13518-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...