Methyl-3-iodo-4-phenoxybenzoate 97%

BD181347-250MG BD181347-1G
BLDPBD181347-250MGEA 62 EUR
BLDPBD181347-250MG BLDPBD181347-1G
Methyl-3-iodo-4-phenoxybenzoate 97%
Methyl-3-iodo-4-phenoxybenzoate

Storage condition: Keep in dark place, Sealed in dry, 2-8°C

Formula: C₁₄H₁₁IO₃
Storage Temperature: Refrigerator
MDL Number: MFCD02912489
CAS Number: 100725-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR