(S)-3-[1-(Dimethylamino)ethyl]phenol 98%

BD18237-100G BD18237-25G
BLDPBD18237-100GEA 825 EUR
BLDPBD18237-100G BLDPBD18237-25G
(S)-3-[1-(Dimethylamino)ethyl]phenol 98%
(S)-3-[1-(Dimethylamino)ethyl]phenol

Storage condition: Keep in dark place, Sealed in dry, Room Temperature

Formula: C₁₀H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD06656491
CAS Number: 139306-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR