Iminodiacetic acid 98%
|
|
BD18318-500G
BD18318-1000G
BLDPBD18318-500GEA
31.5
EUR
BLDPBD18318-500G
BLDPBD18318-1000G
Iminodiacetic acid 98%
Iminodiacetic acid
Storage condition: Keep in dark place, Inert atmosphere, Room temperature
Formula:
C₄H₇NO₄ MW: 133.10 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004280 CAS Number: 142-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...