2-Methyl-1,3-benzothiazole-5-carboxylic acid 95%

BD183359-1G BD183359-250MG
BLDPBD183359-1GEA 66.2 EUR
BLDPBD183359-1G BLDPBD183359-250MG
2-Methyl-1,3-benzothiazole-5-carboxylic acid 95%
2-Methyl-1,3-benzothiazole-5-carboxylic acid

Storage condition: Sealed in dry, Room Temperature

Formula: C₉H₇NO₂S
Storage Temperature: Ambient
MDL Number: MFCD00498510
CAS Number: 24851-69-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR