5-Bromo-3-fluoro-2-hydroxypyridine 98%

BD18606-5G BD18606-10G
BLDPBD18606-5GEA 151 EUR
BLDPBD18606-5G BLDPBD18606-10G
5-Bromo-3-fluoro-2-hydroxypyridine 98%
5-Bromo-3-fluoro-2-hydroxypyridine

Storage condition: Inert atmosphere, Room Temperature

Formula: C₅H₃BrFNO
MW: 191.986 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659498
CAS Number: 156772-63-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR