5-Chloro-2-methyl-1H-indole-3-carbaldehyde 95%
|
|
BD186965-1G
BD186965-250MG
BLDPBD186965-1GEA
259
EUR
BLDPBD186965-1G
BLDPBD186965-250MG
5-Chloro-2-methyl-1H-indole-3-carbaldehyde 95%
5-Chloro-2-methyl-1H-indole-3-carbaldehyde
Storage condition: Inert atmosphere, 2-8°C
Formula:
C₁₀H₈ClNO Storage Temperature: Refrigerator |
MDL Number:
MFCD07186391 CAS Number: 57335-86-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...